SP Wilmad-LabGlass Bruker X Band High Temperature EPR Tube, SP Industries
Supplier: SP Industries
734-QPV-7
76459-978EA 436.54 USD
76459-978
SP Wilmad-LabGlass Bruker X Band High Temperature EPR Tube, SP Industries
Tubes NMR Tubes
This EPR sample tube was made specifically for Bruker’s ER 4114HT-1000 cavity.
  • The ⁷/₁₅ taper joint fits to Wilmad's DWGSK3352 high temperature dewar insert
Three wall finger system made from synthetic quartz yields low thermal expansion coefficient, which results in only a small resonance frequency drift due to wall heating.