SP Wilmad-LabGlass Graded Seal EPR Sample Tube, SP Industries
Supplier: SP Industries
707-GRD-275M
76459-922EA 96.57 USD
76459-922
SP Wilmad-LabGlass Graded Seal EPR Sample Tube, SP Industries
Tubes NMR Tubes
Wilmad's Graded Seal EPR tube is ideal for fusing to an existing valve, glass taper and socket joint.
  • Top of tube is made from low melting point pyrex glass for easy sealing
  • Lower portion of tube is made from high quality clear fused quartz
Rigorous testing is done on each tube.