SP Wilmad-LabGlass Stainless Steel Micro Spatula For NMR and EPR, SP Industries
Supplier: SP Industries
806
76459-580EA 49.55 USD
76459-580
SP Wilmad-LabGlass Stainless Steel Micro Spatula For NMR and EPR, SP Industries
Spatulas
Make solid-state and gel-phase NMR sample trasfers easier.
  • Fits into most 4 mm and larger Ext.Ø. tubes
  • Manufactured from 18/8 stainless steel