HAZWIK® Quick Response Kit, Chemical, Brady Worldwide
Supplier: Brady Worldwide
SKHAZ-QRK-C5
76458-740KT 32.65 USD
76458-740
HAZWIK® Quick Response Kit, Chemical, Brady Worldwide
Spill Control Materials
Get fast and effective spill cleanup with a HAZWIK® Quick response kit. It contains everything you need for emergency spills in a convenient, grab-and-go-style bag.
  • Quickly cleans up hazardous chemical spills
  • Visual instructions on the package
Compact, single-use kit that is ideal for use on small spills in the lab. Contains PPE, tools, absorbent pads, acid and base neutralizers, wipes and disposable materials.
Delivery information: Includes: 6× 7.5×10" pads; 1× 4.5 oz. acid neutralizer; 1× 4.5 oz base neutralizer; 1× pair nitrile gloves; 1× scoop and scrapper; 1× disposal bag; 1× cable tie; 1× surface sanitizing wipe.
Packaging: Sold in packs of 5 kits. Pallet quantity is 250.