Q-sep® Multispeed Centrifuge for QuEChERS, Restek
Supplier: Restek
28295
76458-092EA 3911.66 USD
76458-092
Q-sep® Multispeed Centrifuge for QuEChERS, Restek
Centrifuges Benchtop Centrifuges
The Q-sep centrifuge is a continuous-duty, electronically controlled, horizontal laboratory centrifuge with a lid safety interlock system. The unit is controlled by an electronic push-button timer that is variable from 1 to 30 minutes for precise spin times and ease of use.
  • Samples can be safely viewed through the transparent lid
  • Program 10 custom cycles for time, braking, and speed or g-force
  • Control panel can be temporarily locked on one cycle for error-free reproducibility
  • Cool-flow design prevents samples from overheating by maintaining unit at room
More Product Information
Order Now
SPECIFICATIONS
Motor ½ H.P. Brushless DC
Operating Temperature 16...32 °C
Noise level 64 dB(A)
Power consumption 220 W