LIBERO CS, Single-Use PDF Data Logger, ELPRO
Supplier: ELPRO SERVICES, INC.
800001
76457-552EA 31.39 USD
76457-552
LIBERO CS, Single-Use PDF Data Logger, ELPRO
Data Loggers
The LIBERO CS is the PDF logger that simplifies the shipment process for products with known stability data.
  • Digital display with a visual alarm status that is easy to see, with an 'X' or 'check'
  • All settings, including alarm ranges and logging interval, are simple to program and user-configurable via a free configuration tool
  • High/low or multi-alarm levels with user-defined alarm delay
  • Built-in PDF report generation with embedded raw data (no software required)
This compliant and economic solution for Cold Chain monitoring is ideal for many shipment applications. All settings, including alarm ranges and logging interval, are user-configurable via a free configuration tool. Choose from high/low or multi-alarm levels with user-defined alarm delay. Once configured, insert it into any computer USB port to generate a 21 CFR Part 11 Compliant PDF report with no software or download cables required. Additionally, end users have access to on-screen statistics such as min./max./average, current temperature, and alarm status. If necessary, point-by-point data can be accessed via a free downloadable tool that allows additional notation, reporting, and optional export of raw data to Excel.
Application temperature: Frozen (–20 °C), Cool (2 to 8 °C), refrigerated/ambient (2 to 25 °C), warm (37 to 70 °C).
Certifications: CE marked. The LIBERO CS meets FDA 21 CFR Part 11 and is WHO compliant. The LIBERO CS complies with the ISO standard 19005-1 document management for the long term preservation of electronic documents and is 21 CFR Part 11 and WHO compliant. The CS is delivered with a validation certificate.