VWR® Biology Dissecting Kit with Hinged-Case
Supplier: VWR International
11647-360
76457-538EA 44.35 USD
76457-538
VWR® Biology Dissecting Kit with Hinged-Case
Dissection Sets
Wide variety of VWR® student dissection kits that suit educational needs.
Classroom and research dissecting applications.
Certifications: ISO, GMP
Delivery information: Includes hinged case with hard plastic snap cover and compartments for 12 instruments: #4 surgical handle; #22 scalpel blades (6); 6" narrow blade dissecting scalpel: standard 1.5" cut edge; 5-¹/₂" sharp/sharp scissors; 4-¹/₂" straight iris scissors; 5-¹/₂" straight thumb forceps: round, serrated tips, serrated grip; 4-¹/₂" curved forceps: medium serrated tips, serrated grip; Stainless steel mall-probe and seeker: hexagon 3³/₄" handle, 2¹/₂" seeker; Straight teasing needle with 4" plastic handle; Angular teasing needle with 4" plastic handle; Glass dropper with rubber bulb, large size 5"; 6" hard plastic ruler.