VWR® University High-Grade Dissecting Kit
Supplier: VWR International
76457-534
76457-534EA 66.51 USD
76457-534
VWR® University High-Grade Dissecting Kit
Dissection Sets
Wide variety of VWR® student dissection kits that suit educational needs.
Classroom and research dissecting applications.
Certifications: ISO, GMP
Delivery information: Includes black zippered case with velvet liner, 4-¹/₂" straight iris scissors; 5-¹/₂" sharp/blunt operating scissors; 4-¹/₂" straight forceps (medium serrated tips, serrated grip with guide pin); 4-¹/₂" straight 1×2 teeth tissue forceps; 5-¹/₂" stainless steel Kelly haemostat (curved); #3 scalpel handle; #10 scalpel blades (pk 10); 65" cartilage knife (2" cutting edge); stainless steel mall-probe and seeker (hexagon 3³/₄" handle; 2¹/₂" seeker); straight teasing needle with 325" steel adjustable chuck holder; bent teasing needle with 325" steel adjustable chuck holder; 6"" hard plastic ruler.