VWR® Elementary Dissecting Kit - Vinyl Case
Supplier: VWR International
11647-354
76457-526EA 22.64 USD
76457-526
VWR® Elementary Dissecting Kit - Vinyl Case
Dissection Sets
Wide variety of VWR® student dissection kits that suit educational needs.
Classroom and research dissecting applications.
Certifications: ISO, GMP
Delivery information: Includes single fold vinyl case with lining, 6" student dissecting scalpel (screw-lock with blade); 45" student dissecting scissors (sharp point); 45" straight forceps (wide thumb-grip, round tip); straight teasing needle with 4" plastic handle; angular teasing needle with 4" plastic handle; glass dropper with rubber bulb (large 5" size); 6" hard plastic ruler.