VWR® Zippy Dissecting Kit with Scalpel
Supplier: VWR International
11647-352
76457-522EA 22.37 USD
76457-522
VWR® Zippy Dissecting Kit with Scalpel
Dissection Sets
Wide variety of VWR® student dissection kits that suit educational needs.
Classroom and research dissecting applications.
Certifications: ISO, GMP
Delivery information: Includes zippered case with 6" student dissecting scalpel (standard 15" cut edge); 45" student dissecting scissors (sharp point); 45" student forceps (blunt, serrated tips; serrated grip); straight teasing needle with 4" plastic handle; angular teasing needle with 4" plastic handle; glass dropper with rubber bulb (large 5" size); 6" hard plastic ruler.