VWR® Premium Metzenbaum Dissecting Scissors, German Steel
Supplier: VWR International
11647-264
76457-346EA 57.22 USD
76457-346
VWR® Premium Metzenbaum Dissecting Scissors, German Steel
Scissors
These high-quality German steel dissecting scissors are versatile and can be used in multiple procedures and settings
  • Large rings for comfort, user control and stability
  • Manufactured from ASTM-standard German stainless steel
  • Anti-glare coating
  • Autoclavable and corrosion-resistant
Top-tier instrument with superior craftsmanship. Product conforms to international quality standards.
Certifications: ISO, GMP