HumaDerm Human Skin Collagen Type I, 6 mg/ml Solution, Humabiologics
Supplier: HUMABIOLOGICS, INC
SCS6-10ML
76452-620EA 410.56 USD
76452-620
HumaDerm Human Skin Collagen Type I, 6 mg/ml Solution, Humabiologics
Cell Culture Supplements and Additives
Clinically relevant native human skin collagen for hydrogel and bioink applications.
  • Native Human Skin Collagen
  • Clinically relevant and xeno-free for regenerative medicine applications
  • 3D hydrogel or bioink
  • Supports various cell attachments
HumaDerm is isolated from human skin sourced strictly from tissue partners who comply with the requirements for transplantable human tissue. HumaDerm has undergone a proprietary manufacturing method to preserve the native human collagen found in skin. HumaDerm Type I Collagen is supplied at 6 mg/ml for hydrogel and bio-ink applications, and can be diluted for custom applications.
HumaDerm addresses the growing need for more clinically relevant and animal-free human tissue constructs and models. Each lot of HumaDerm has undergone extensive testing to meet high safety and quality requirements for translational research needs.
All tissue has been tested for safety and found negative for communicable diseases and infectious agents.
Ordering information: HumaDerm is available at 6 mg/ml in 10 ml.
Caution: HumaDerm is for research use only and is not intended for human use, diagnosis, screening, household, food or other uses.