HumaCoat Human Skin Collagen Type I, 1 mg/ml Solution, Humabiologics
Supplier: HUMABIOLOGICS, INC
SCS1-10ML
76452-614EA 153.95 USD
76452-614
HumaCoat Human Skin Collagen Type I, 1 mg/ml Solution, Humabiologics
Cell Culture Supplements and Additives
Clinically relevant native human skin collagen for cell culture coating applications.
  • Native Human Skin Collagen for thin coating
  • Clinically relevant and xeno-free for regenerative medicine applications
  • Ready to use
  • Supports various cell attachments
HumaCoat is isolated from human skin sourced strictly from tissue partners who comply with the requirements for transplantable human tissue. HumaCoat has undergone a proprietary manufacturing method to preserve the native human collagen found in skin. HumaCoat Type I Collagen is supplied at 1 mg/ml for cell culture applications and can be diluted for optimal desired concentration.
HumaCoat addresses the growing need for more clinically relevant and animal-free human tissue constructs and models. Each lot of HumaCoat has undergone extensive testing to meet high safety and quality requirements for translational research needs.
All tissue has been tested for safety and found negative for communicable diseases and infectious agents.
Ordering information: HumaCoat is available at 1 mg/ml in 10 ml.
Caution: HumaCoat is for research use only and is not intended for human use, diagnosis, screening, household, food or other uses.