exsig™ Mag Ultra-Sensitive Magnetic Bead Based Extraction System, Primerdesign
Supplier: PRIMER DESIGN LTD
R35000
76452-342EA 28288.28 USD
76452-342
exsig™ Mag Ultra-Sensitive Magnetic Bead Based Extraction System, Primerdesign
Automated Nucleic Acid Purification Systems
exsig™ Mag facilitates a high-quality, high-throughput workflow solution by providing flexible extraction chemistry through its compatibility with widely used liquid handlers and extraction robots.
  • Ultra-sensitive nucleic acid purification
  • 5000 extractions
  • Fully automatable
  • Flexible chemistry compatible with a wide variety of sample types
  • Open platform that can be deployed on programmable extraction robotics
It compliments the genesig® real-time PCR COVID-19.