Falcon® Pipette Controller, Corning
Supplier: Corning
357469
10028-602CS 730.31 USD
10028-602 76451-998
Falcon® Pipette Controller, Corning
Pipettes Pipette Controllers
This light, ergonomic, motorized pipette controller is designed for use with glass and plastic serological pipettes from 0.5 to 100 ml.
  • Ergonomic design for exceptional comfort
  • Nosepiece with autoclavable pipette holder and filter
  • Ergonomically positioned finger triggers provide comfortable grip
  • Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and viscosities
  • LCD display for continuous visualization of speed, modes, and battery status
  • Easy access to the battery compartment
  • Filters, pipette holder and nosepiece are autoclavable
Delivery information: Supplied with a two-position charging stand, a universal battery charger, three batteries, two 0.2 μm and two 0.45 μm hydrophobic polytetrafluoroethylene (PTFE) filters.