VWR® Cleanroom Mask with Earloops, Made in USA
Supplier: VWR International
85002
76451-956CS 168.1 USD
76451-956
VWR® Cleanroom Mask with Earloops, Made in USA
Masks Cleanroom Masks
The combination of strong BFE and PFE make these masks ideal for use in production or research areas.
  • Protection against bacteria and particles
  • 3-ply construction
  • Non sterile
  • With earloops
Part of the VWR® Made in USA PPE line, these cleanroom masks with earloops are a core item of any solid garment program, offering the adjustability user's desire.
Country of origin: USA.
Ordering information: Made in USA Masks with ties are also available; see catalog number 76409-318.
Packaging: Masks are packaged 50 per bag, 10 bags per case.