Keystone CRP Gamma Irradiated Cleanroom Trash Bags, 2 Mil Thick, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
S-3655CP-1 S3655CP1DP S3655CP2 S3655CP2DP S3655CP3 S3655CP3DP S3655CP4 S3655CP4DP S3655CP5 S3655CP5DP S-3655CP-10 S3655CP10DP S-2428CP-1 S2428CP10 S2428CP10DP S2428CP1DP S-2428CP-5 S2428CP5DP
76451-814CS 849.75 USD
76451-814 76451-816 76451-818 76451-820 76451-822 76451-824 76451-826 76451-828 76451-830 76451-832 76451-834 76451-836 76451-838 76451-840 76451-842 76451-844 76451-846 76451-848
Keystone CRP Gamma Irradiated Cleanroom Trash Bags, 2 Mil Thick, Keystone Cleanroom Products
Bags Waste Disposal Bags
Ready to use pre-sterilized trash bags make it easy to remove waste from pharmaceutical production areas without introducing potential sources of contamination.
  • Gamma irradiated
  • Convenient way to hold refuse or other items for removal from an aseptic production area
These rugged bags are supplied sterile in easy-to-open Tyvek® Peel pouches and eliminate the need to sterilize or sanitize waste containers or bags prior to bringing them into the controlled environment. Each pouch includes a lot number for traceability.
Certifications: Certificates of Irradiation and Conformance with each shipment.