DuPont™ Tyvek® 1073B, Autoclave Wrappers, Round, Keystone Cleanroom Products
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
MGTVKSHEET0606 MGTVKSHEET10 MGTVKSHEET14
76450-766CS 2536.79 USD
76450-766 76450-784 76450-792
DuPont™ Tyvek® 1073B, Autoclave Wrappers, Round, Keystone Cleanroom Products
Covers
Improve the microbial barrier and cleanliness of your sterilization wrap.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Round shaped Tyvek® Wrappers are an ideal way to cover round openings for steam sterilization. Eliminate the wasted time and expense of cutting larger square sheets of sterilization wrap to desired shapes and sizes. Clean cut circles delivered in poly bags ensure cleanliness. Simply secure in place with autoclaveable elastic bands.
Delivery information: Certificates of conformance sent with each shipment.