RAPIDPoint® 500e Blood Gas Analyzer, Siemens Healthineers
Supplier: SIEMENS
11416751
76449-582EA 57720.05 USD
76449-582
RAPIDPoint® 500e Blood Gas Analyzer, Siemens Healthineers
Clinical Diagnostic Supplies Clinical Analysers
The RAPIDPoint® 500e system elevates Siemens Healthineers blood gas offering to a new level, allowing you to spend more time focused on patient care. Implement a proven end-to-end blood gas solution that reduces the daily burden of device management and enables you to give your full care and attention where it matters most. With an improved user experience, the analyzer transforms care delivery, setting an elevated standard in simplicity, quality assurance, and data security for healthcare organizations.
  • Comprehensive menu to help clinical decision making
  • Maintenance-free cartridge-based technology
  • Full test menu from a single 100 µL blood sample
Integri-sense Technology:
- Automatic QC ensures instrument is running correctly
- Clot management and intelligent calibration ensures sample integrity
- Establish critical ranges across all sample types and electronically document critical values
Enhanced data security:
- Windows 10 Operating System
- Encrypted data transmission, 2-step authentication, anti-malware and firewall
- Anytime software update (ASU)