VWR® Adhesive Sealing Strips, PP, 1×8
Supplier: VWR International
76449-282 76449-284
76449-282CS 28.19 USD
76449-282 76449-284
VWR® Adhesive Sealing Strips, PP, 1×8
Seals Microplate Seals
1x8 adhesive sealing strips are ideal for use with 1x8 strip-well ELISA plates.
  • Ideal for ELISA, incubation and general use
  • Seals a single 8-well row of a 96-well plate or an ELISA strip-well plate
  • 2.0 mil PP film
  • Compatible with both polystyrene and polypropylene plates
  • Certified DNase- and RNase-free
  • Recommended temperature range from −40 to +120 °C
8 strips are precut on a continuous liner for ease of use.