VWR® Adhesive Sealing Strips for 8-Strip PCR Tubes
Supplier: VWR International
76449-280
76449-280CS 47.23 USD
76449-280
VWR® Adhesive Sealing Strips for 8-Strip PCR Tubes
Seals Microplate Seals
Easier to apply and remove as compared to standard molded PCR strip caps.
  • Ideal for PCR and storage
  • Alternative to strip-caps; no sore thumbs and fingers guaranteed
  • Seals a single 8-well row of a 96-well plate or a single PCR 8-tube strip
  • Certified DNase- and RNase-free
  • 2.0 mil transparent PE film
  • Recommended temperature range from −20 to +120 °C
8 strips are precut on a continuous liner for ease of use.