mRNA Decapping
Supplier: New England Biolabs (NEB)
M0608S
76447-140EA 112 USD
76447-140
mRNA Decapping
Enzymes
mRNA Decapping Enzyme decaps m7G-capped mRNA producing a 5′ monophosphate end.
  • Efficient replacement for Tobacco Acid Pyrophosphatase
  • Cap0 and Cap1 are removed with equal efficiency
  • Suitable for 5′ RLM-RACE and RNA-seq
  • 1 µl will decap 20 µg of a 1.7 kb mRNA in 15 minutes