VWR® PCR Plate Centrifuge II
Supplier: VWR International
76447-106
76447-106EA 693.41 USD
76447-106
VWR® PCR Plate Centrifuge II
Centrifuges Microcentrifuges
Quickly spins down droplets in PCR plates maximizing PCR yield.
  • Compact size
  • Built-in timer and display
  • Bushless DC motor
  • Electronic braking system
  • Accepts all popular PCR plates
Centrifugation of microplates before PCR ensures all reactants are in the bottom of the wells for optimized concentrations and improved yields. Plates are inserted vertically and quickly spin to a maximum speed of 2800 rpm (580×g). Most samples can be spun down quickly, in less than 20 seconds.
Certifications: CE marked