Linearity FLQ Anemia Siemens Centaur, AUDIT Microcontrols
Supplier: AUDIT MICROCONTROLS
K819M-5
76446-936EA 356.45 USD
76446-936
Linearity FLQ Anemia Siemens Centaur, AUDIT Microcontrols
Controls and Calibration Verification Materials
Linearity FLQ anemia for Siemens centaur is an IVD device consisting of five levels.
  • Format: frozen liquid
  • 5 levels
It is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: cortisol, ferritin, folate, vitamin B12.