Linearity FLQ TDM for Roche, AUDIT Microcontrols
Supplier: AUDIT MICROCONTROLS
K933M-5
76446-934EA 487.51 USD
76446-934
Linearity FLQ TDM for Roche, AUDIT Microcontrols
Controls and Calibration Verification Materials
Linearity FLQ TDM for Roche systems is an IVD device consisting of five levels.
  • Format: Frozen liquid
It is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: acetominophen, amikacin, carbamezepine. digoxin, gentamicin, NAPA, phenobarbital, phenytoin, proccainamide, quinidine, salicylate, theophylline, tobramycin, valproic acid, vancomycin.