Mid-Turbinate Adult Flocked Swab With Stopper, C&A Scientific
Supplier: Hardy Diagnostics
972014
76446-898EA 97.36 USD
76446-898
Mid-Turbinate Adult Flocked Swab With Stopper, C&A Scientific
Swabs Swabs for Clinical Use
Hardy Diagnostics offers premium flocked swabs.
  • Highly absorbent tip
  • Soft smooth, non-abrasive
Our swabs ensure maximum collection and release of specimens, specifically for applications in relation to COVID-19. With integrated 80mm breakpoint on our flocked swab, it is easily paired with our Viral Transport Medium (Cat. no. 76334-640) for the preservation of viral specimens.