Modified Trichrome Blue Stain for Microsporidia, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
MT016
76446-784EA
262.19
USD
76446-784
Modified Trichrome Blue Stain for Microsporidia, Hardy Diagnostics
Stains
Microbiology Stains
For staining microsporidia, formulated according to Weber and Ryan, with aniline blue.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)