TRIO.BAS™ Air Sampler Software PC
Supplier: Hardy Diagnostics
BAS296
76446-766EA 3150.54 USD
76446-766
TRIO.BAS™ Air Sampler Software PC
Software Laboratory Software
BAS Software allows the transfer of environmental sampling data from a bluetooth or data transfer cable equipped TRIO.BAS™ instrument to and from a personal computer. The software allows database management, editing, reporting, and remote control of the instrument.
  • Management of users, passwords, and related permissions
  • Communication and data transfer between TRIO.BAS™ air samplers
  • Collected data cannot be modified according to CFR
  • ALCOA compliant
  • Permits input of other environmental sampling information e.g: surface, passive air sampling (settling plates),
More Product Information