pZerve DMSO Free Cryopreservation Solutions, Protide Pharmaceuticals
Supplier: PROTIDE PHARMACEUTICALS
5720
5700
76443-300EA
120.1
USD
76443-300
76443-302
pZerve DMSO Free Cryopreservation Solutions, Protide Pharmaceuticals
Cell Culture Supplements and Additives
A unique cryopreservation solution that does not contain DMSO, fetal bovine serum, or animal protein. pZerve™ is ready to use and does not require any dilution or further processing.
- Does not contain DMSO
- No fetal bovine serum or other animal proteins
- Sterile/Ready to use
Certifications: cGMP manufactured.