TCM Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Supplier: PROTIDE PHARMACEUTICALS
1001 1002 1005
76443-252EA 122.14 USD
76443-252 76443-254 76443-256
TCM Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Cell Culture Supplements and Additives
General serum replacement for many cell types from a variety of species. TCM™ is a defined, multi-purpose serum replacement useful in studying the effects of growth factors and the production of proteins. TCM™ contains highly purified, heat-treated bovine serum albumin, heat-treated bovine transferrin and recombinant insulin.
  • Defined, serum-free
  • No hormones, antibiotics, growth factors, phenol red, Ig
  • 50X concentrate
  • Can be utilized with a wide variety of basal media
Certifications: cGMP manufactured.