PLET Agar, for Bacillus anthracis
Supplier: HARDY DIAGNOSTICS
G153
76443-248PK 62.81 USD
76443-248
PLET Agar, for Bacillus anthracis
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of Bacillus anthracis from environmental samples, animal products, carcasses, and clinical samples from non-sterile sites.