DP Series Industrial Vacuum Drying Oven, Yamato Scientific
Supplier: Yamato Scientific
DP-83C
76443-174EA 30519.22 USD
76443-174 76443-176
DP Series Industrial Vacuum Drying Oven, Yamato Scientific
Ovens Vacuum Ovens
Large scale vacuum drying oven designed for treatment of large-sized parts.
  • Vacuum pump can be installed inside the oven
  • Stainless steel plate interior, NW40 flange air exhaust port
  • Decompressed chamber wall direct heating method
  • Glass fiber insulating material
  • SSR driving heater circuit control
  • One year warranty
Quick connect/disconnect of vacuum pipes for easy vacuum pump maintenance. Because the exhaust system is improved to significantly shorten the time to reach vacuum, working efficiency is improved. Use specialized function menu key and up and down key to set. With program operation function, use submenu key to operate overheat protector, deviation correction,and more.
Safety device includes self-diagnosis circuit (abnormal temp. sensing, heater disconnection, auto overheat prevention, SSR short circuit), overheat protector, EBL to prevent overcurrent, key lock.
Ordering information: Stainless steel punching plates. Shelf plate, vacuum pump, N₂ introduction device, recorder, alarm indicator lamp (stand-by/running/malfunction), external communication (RS485), temp. output terminal (4 to 20 mA), Output terminal for external alarm, time up output terminal (not included).