Digital Oil Baths for Rotary Evaporator, 200 - 230 V, Yamato
Supplier: Yamato Scientific
BO-312A
76443-168EA 938.15 USD
76443-168
Digital Oil Baths for Rotary Evaporator, 200 - 230 V, Yamato
Baths Oil Baths
These digital oil baths are easy to use, they offer digital settingin a compact design oil bath.
  • 200 - 230 V model, not for use in USA
  • Fixed temperature operation function
  • White LED digital display
  • One year warranty
Exclusive connection for bath operational setting function with RE601/801 rotary evaporator, removable oil tank for convenient cleaning and changing of oil (75846-372). Flat-shaped bath with no heater or sensor inside makes for ease in cleaning (76443-166, 76443-168).
Safety device includes bath protection sheet, automatic overheat prevention, independent overheat prevention (fixed temperature), temperature upper limit error, overcurrent protection fuse; overshoot alert, auto resume (selectable), 2A service outlet, calibration offset (76443-166, 76443-168). Bath protection cover (ABS heat-resistant resin), self-diagnostic function (automatic overheat prevention, temperature sensor error, triac short circuit, heater disconnection, main relay failure), circuit protector, thermal fuse, micro switch to detect heating without oil (75846-372).
Caution: 200 - 230 V model without plug, not for use in USA. Corresponding model with voltage suitable for use in US is also available, please see separate product page.