ThermoChill II Recirculating Chiller, PD-1 Pump, Thermo Scientific
Supplier: Thermo Fisher Scientific
197211010000
76443-110EA 5620 USD
76443-110
ThermoChill II Recirculating Chiller, PD-1 Pump, Thermo Scientific
Circulating Thermostats Closed Circulator Systems Cooling Circulators
ThermoChill II provides reliable liquid temperature control, with cooling capacity up to 1000 w/3415 BTU.
  • Accessible fill port for easy filling
  • Pressure gauge located on control panel for easy viewing
  • Intuitive digital controller
  • Controller indicator lights illuminate when adjusting settings or when high/low temp alarms activated
  • Removable full-low strainer ensures clean recirculating fluid
  • Locking caster for easy unit maneuvering
  • RS232 or RS485 digital communications optional
Thermo Scientific™ ThermoChill II Recirculating Chillers are a compact line of refrigerated recirculators ranging in cooling capacities from 600 to 2000 watts. Innovative design features such as minimized plumbing connections and an integrated full-flow fluid strainer maximize product reliability. With simplicity in design and operation, the ThermoChill platform will provide years of reliable liquid temperature control to meet all of your conventional application requirements.