Legacy Everwipe Disinfectant Wipes, Essendant
Supplier: Janitorial Supplies
LEY10100
76442-800CT 235.79 USD
76442-800
Legacy Everwipe Disinfectant Wipes, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
The Everwipe Disinfectant Wipe is an EPA registered, hospital-grade disinfectant that quickly and effectively disinfects hard, non-pourous surfaces. Kills bacteria, viruses, and fungi.
  • Cleans, disinfects and deodorizes
  • Alchohol free, bleach free
  • Quat based formulation
  • Application: General purpose
Applicable material: Hard, non-porous, inanimate environmental surfaces; floors; walls; metal surfaces; stainless steel; glazed porcelain; glazed ceramic tile; plastic; bathroom shower stalls.