Intelsius® Biomailer, DGP Intelsius
Supplier: DGP INTELSIUS LLC
BM003
76441-380CS 423.52 USD
76441-380
Intelsius® Biomailer, DGP Intelsius
Mailers and Shippers
The Intelsius® Biomailer is designed for use within national and global postal networks shipping small specimens, such as blood vials and slides.
  • Rigid plastic case
  • Ship small specimens
  • National and global postal mailer
  • Supplied with plastic pouch and absorbent
  • Multi-use device
The rigid plastic case ensures that samples are protected during transit, and allows the system to be refurbished and used again, offering both a cost saving and environmental benefit.
Certifications: ISTA Certified Class 6.2, Category B.
Ordering information: Includes A5 Pathopouch (95 kPa secondary), 4 bay absorbent, protective sleeve, UN3373 Biological Substance Category B label, and security seal label.