Caffeic Acid Disks, for Cryptococcus, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z118
76441-346EA 84.43 USD
76441-346
Caffeic Acid Disks, for Cryptococcus, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
HardyDisk™ Caffeic Acid Disks detect the ability of an organism to produce the enzyme phenyloxidase, which is useful for the identification of Cryptococcus neoformans and Cryptococcus gatti.
  • Detect the ability of an organism to produce the enzyme phenyloxidase
  • Useful for the identification of Cryptococcus neoformans and Cryptococcus gattii