High Performance FR Long Sleeve Knit Henley, Khaki, Lakeland Industries
Supplier: Lakeland
HSAT20-2X HSAT20-2XT HSAT20-3X HSAT20-3XT HSAT20-4X HSAT20-4XT HSAT20-5X HSAT20-6X HSAT20-LG HSAT20-LGT HSAT20-MD HSAT20-MDT HSAT20-SM HSAT20-XL HSAT20-XLT HSAT20-XS
76437-572EA 162.34 USD
76437-572 76437-574 76437-576 76437-578 76437-580 76437-582 76437-584 76437-586 76437-588 76437-590 76437-592 76437-594 76437-596 76437-598 76437-600 76437-602
High Performance FR Long Sleeve Knit Henley, Khaki, Lakeland Industries
Workwear
Lightweight, inherently FR Long Sleeve Knit Henley with permanent moisture wicking properties make this the most comfortable FR/AR base layer.
  • Unparalleled CAT 2 protection at 16.4 cal/cm²
  • Lightweight 6.95 oz. inherently FR fabric
  • Excellent breathability
  • Permanent moisture wicking properties
  • Tag free collar
  • Drop shoulder design
  • Left chest pocket
BASE layers are the foundational key to the Lakeland FR/AR Layering System. To be cool during hot summer days, and warm when it's frigid cold outside, wearers need to be dry. BASE layer shirts have the optimal 60/40 blend of hydrophobic and hydrophilic fibers to constantly pull moisture away from the body, move it to the outer surface of the garment, and allow it to dry. Staying dry is the key to keeping wearers comfortable and better protected.
Certifications: NFPA 70E. NFPA 2112 - UL certified. ATPV: 16.4 cal/cm². CAT: 2.