Alethia® HSV 1 and 2, Meridian Bioscience
Supplier: MERIDIAN DIAGNOSTICS
480650
76437-458EA 3118.56 USD
76437-458
Alethia® HSV 1 and 2, Meridian Bioscience
Assays Immunological Assays
Qualitative in vitro diagnostic test for the direct detection and differentiation of herpes simplex virus type 1 (HSV-1) and herpes simplex virus type 2 (HSV-2) DNA in cutaneous and mucocutaneous lesion specimens from male and female patients suspected of Herpetic lesions.
  • Alethia® HSV 1 and 2 offers significant advantages over traditional viral culture by providing same day results from both cutaneous and mucocutaneous specimen types
  • Alethia® is a fast, accurate and reliable molecular solution that is easily adaptable in any laboratory with minimal hands-on time and no specialized training
  • Alethia® HSV 1 and 2 provides an actionable result to enable physicians to quickly start treatment of patients
  • The Alethia® HSV 1 and 2 is a sophisticated molecular test which can also help pediatricians educate families of newborn babies about the risks and dangers of HSV 1 and 2