Alethia® Group B Streptococcus, Meridian Bioscience
Supplier: MERIDIAN DIAGNOSTICS
480350
76437-456EA 6745.65 USD
76437-456
Alethia® Group B Streptococcus, Meridian Bioscience
Assays Immunological Assays
The Alethia® Group B Streptococcus assay is a qualitative in vitro diagnostic for the detection of Streptococcus agalactiae in enriched cultures obtained from vaginal/rectal swab specimens from antepartum women.