Alethia® C. difficile DNA amplification assay, Meridian Bioscience
Supplier: MERIDIAN DIAGNOSTICS
480050
76437-450EA 6745.65 USD
76437-450
Alethia® C. difficile DNA amplification assay, Meridian Bioscience
Assays Immunological Assays
The Alethia C. difficile DNA amplification assay, performed on the Alethia reader, is a qualitative in-vitro diagnostic test for the direct detection of toxigenic C. difficile in human stool specimens from pediatric and adult patients suspected of having Clostridium difficile-associated disease (CDAD).
  • Reduce the risk of missing a true positive with the accuracy of molecular
Fast, actionable results enable physicians to make informed decisions about the patient management and treatment resulting in reduced healthcare costs associated with isolation and antibiotics.