Immunocard® Mycoplasma, Meridian Bioscience
Supplier: MERIDIAN DIAGNOSTICS
709030
76437-438EA 1565.99 USD
76437-438
Immunocard® Mycoplasma, Meridian Bioscience
Assays Immunological Assays
The ImmunoCard Mycoplasma enzyme immunoassay (EIA) is an in vitro qualitative procedure for the detection of IgM to Mycoplasma pneumoniae in human serum. Test results are intended to aid in the diagnosis of recent Mycoplasma pneumoniae infection.