Immunocard STAT!® Strep A, Meridian
Supplier: MERIDIAN DIAGNOSTICS
755250
76437-434EA 933.13 USD
76437-434
Immunocard STAT!® Strep A, Meridian
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Alethia Group A Streptococcus assay, is a qualitative invitro diagnostic test for the detection of Streptococcus pyogenes (Group A β-hemolytic Streptococcus) in throat swab specimens.
  • CLIA Waived, easy to use technology
  • Rapid test for detection of Group A Streptococcal Antigen from throat swabs
  • Results in five minutes, shortening patient wait time
  • Test with Immunocard STAT! Strep A first and be able to treat while patient is waiting
  • Reflex Rapid Group A Strep negatives to the Alethia Group A Strep molecular to confirm results