Temperature Probes, Environmental Express®
Supplier: Environmental Express
SC980 SC981 SC985
76437-122EA 282.54 USD
76437-122 76437-124 76437-126
Temperature Probes, Environmental Express®
Temperature Probes
Temperature probes enhance usage of Hot Blocks.
  • Compatible with Environmental Express® HotBlock® systems
The heads-up Lollipop™ Traceable® thermometer allows technicians to monitor actual sample temperature in 50 mL samples. The PTFE holder sits on the top of the digestion vessel. Thermometer has LCD display and can be set in Fahrenheit or Celsius. The stem is coated with PTFE to help in corrosive environments. Wide range covers 99 percent of the lab's routine temperature measurements. Solid-state construction eliminates dangers inherent in breakable glass and toxic mercury thermometers.
Delivery information: Kit contains the holder ring and thermometer with NIST Certification. NIST calibration good for at least 1 year from date of shipment.