Sodium hydroxide concentrate 1.25 N, SimpleDist Snip & Pour™ Reagent
Supplier: ENVIRONMENTAL EXPRESS
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
C7115
76436-850EA
105
USD
76436-850
Sodium hydroxide concentrate 1.25 N, SimpleDist Snip & Pour™ Reagent
Sodium hydroxide
Deliver precise volume with pre-measured reagent.
- Easy-to-use, just snip and pour into receiver tube
- Increase accuracy by eliminating the need to measure and pipette
Snip & Pour Concentrates are packaged in convenient PTFE tubes. Reagent addition is a snap with Snip & Pour reagent tubes from Environmental Express. Just snip the corner of the PTFE tube and pour the contents directly into the collection trap (SimpleDistTM). Each pre-measured PTFE tube for SimpleDist delivers 5 ml of reagent.
Formula:
NaOH |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...