TKN Tubes, Environmental Express®
Supplier: Environmental Express
C6010
76436-824EA 112.47 USD
76436-824
TKN Tubes, Environmental Express®
Tubes Test Tubes
Optimize your TKN analysis for better results.
  • Interchangeable glassware works with SimpleDist® and TKN HotBLock® systems
  • Maximum heat transfer with flat-bottom, borosilicate glassware
  • Ideal for reflux and digestion methods
Tube features 30200 mm dimensions with 38 mm closure without cap and graduated at 50 ml mark.