Exel International Bulk, Unsterile Syringes, Air-Tite Products
Supplier: Air-Tite
EB5LL
76436-742CS 451.13 USD
76436-742
Exel International Bulk, Unsterile Syringes, Air-Tite Products
Syringes General-Purpose Syringes
The bulk, unsterile version of EXEL International's quality syringes. EXEL is known for unsurpassed quality disposable syringes. Great value at resonable costs.
  • Standard 3-part syringe
  • Latex- and pyrogen-free
  • Non-toxic
  • Positive plunger stop
  • Disposable
EXEL International is known for unsurpassed quality at a low price. The design includes bold print, clear barrels, and siliconized gaskets for smooth, consistent plunger motion and a positive plunger stop.
Certifications: Meets ISO standards.
Ordering information: Needles available separately.