Phosphate Buffered Saline (PBS) with 0.05% Tween® 20, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
U235
76436-460PK 167.25 USD
76436-460
Phosphate Buffered Saline (PBS) with 0.05% Tween® 20, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Used during specimen processing for recovery of bacterial endospores.
Recommended as a component of anthrax screening programs for use in the nasal swab heat-shock procedure to recover Bacillus anthracis endospores.