Avantor® Hichrom HI-SE54, GC Columns
Supplier: VWR International
76430-914 76430-916 76430-918 76430-920 76430-922 76430-924 76430-926 76430-928 76430-930 76430-932 76430-934 76430-936 76430-938 76430-940 76430-942 76430-944 76430-946 76431-404 76431-406 76431-408 76431-410 76431-412 76431-414 76431-416 76431-418 76431-420 76435-472 76435-474 76435-476 76435-478 76435-480 76435-482 76435-484 76435-486 76435-488 76435-490 76435-492 76435-494 76435-496 76435-498 76435-500 76435-502 76435-504 76435-506 76435-508 76435-510 76435-512 76435-514 76435-516 76435-518 76435-520 76435-522 76435-524 76435-526 76435-528 76435-530 76435-532 76435-534 76435-536 76435-538 76435-540 76435-542 76435-544 76435-546 76435-548 76435-550 76435-552 76435-554 76435-556 76435-558 76435-560 76435-562 76435-564 76435-566 76435-568 76435-570 76435-572 76435-574 76435-576 76435-578 76435-580 76435-582 76435-584 76435-586 76435-588 76435-590 76435-592 76435-594 76435-596 76435-598 76435-600 76435-602 76435-604 76435-606 76435-608 76435-610 76435-612 76435-614 76435-616 76435-618 76435-620 76435-622 76435-624 76435-626 76435-628 76435-630 76435-632 76435-634 76435-636 76435-638 76435-640 76435-642 76435-644 76435-646 76435-648 76435-650 76435-652 76435-654 76435-656 76435-658 76435-660 76435-662 76435-664 76435-666 76435-668 76435-670 76435-672 76435-674 76435-676 76435-678 76435-680 76435-682 76435-684 76435-686 76435-688 76435-690 76435-692 76435-694 76435-696 76435-698 76435-700 76435-702 76435-704 76435-706 76435-708 76435-710 76435-712 76435-714 76435-716 76435-718 76435-720 76435-722 76435-724 76435-726 76435-728 76435-730 76435-732 76435-734 76435-736 76435-738 76435-740 76435-742 76435-744 76435-746 76435-748
76430-914EA 910.86 USD
76430-914 76430-916 76430-918 76430-920 76430-922 76430-924 76430-926 76430-928 76430-930 76430-932 76430-934 76430-936 76430-938 76430-940 76430-942 76430-944 76430-946 76431-404 76431-406 76431-408 76431-410 76431-412 76431-414 76431-416 76431-418 76431-420 76435-472 76435-474 76435-476 76435-478 76435-480 76435-482 76435-484 76435-486 76435-488 76435-490 76435-492 76435-494 76435-496 76435-498 76435-500 76435-502 76435-504 76435-506 76435-508 76435-510 76435-512 76435-514 76435-516 76435-518 76435-520 76435-522 76435-524 76435-526 76435-528 76435-530 76435-532 76435-534 76435-536 76435-538 76435-540 76435-542 76435-544 76435-546 76435-548 76435-550 76435-552 76435-554 76435-556 76435-558 76435-560 76435-562 76435-564 76435-566 76435-568 76435-570 76435-572 76435-574 76435-576 76435-578 76435-580 76435-582 76435-584 76435-586 76435-588 76435-590 76435-592 76435-594 76435-596 76435-598 76435-600 76435-602 76435-604 76435-606 76435-608 76435-610 76435-612 76435-614 76435-616 76435-618 76435-620 76435-622 76435-624 76435-626 76435-628 76435-630 76435-632 76435-634 76435-636 76435-638 76435-640 76435-642 76435-644 76435-646 76435-648 76435-650 76435-652 76435-654 76435-656 76435-658 76435-660 76435-662 76435-664 76435-666 76435-668 76435-670 76435-672 76435-674 76435-676 76435-678 76435-680 76435-682 76435-684 76435-686 76435-688 76435-690 76435-692 76435-694 76435-696 76435-698 76435-700 76435-702 76435-704 76435-706 76435-708 76435-710 76435-712 76435-714 76435-716 76435-718 76435-720 76435-722 76435-724 76435-726 76435-728 76435-730 76435-732 76435-734 76435-736 76435-738 76435-740 76435-742 76435-744 76435-746 76435-748
Avantor® Hichrom HI-SE54, GC Columns
Chromatography Columns
General purpose column for solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, allergens, pesticides, and so on.
Max temperature limits may change depending on the film thickness.