Avantor® Hichrom HI-35 MS, GC Columns
Supplier: VWR International
76431-148 76431-150 76431-152 76431-154 76431-156 76431-158 76431-160 76431-162 76431-164 76431-166 76431-168 76431-170 76432-398 76432-400 76432-402 76432-404 76432-406 76432-408 76432-410 76432-412 76432-414 76432-416 76432-418 76432-420 76432-422 76432-424 76432-426 76432-428 76432-430 76432-432 76432-434 76432-436 76432-438 76432-440 76432-442 76432-444 76432-446 76432-448 76432-450 76432-452 76432-454 76432-456 76432-458 76432-460 76432-462 76432-464 76432-466 76432-468 76432-470 76432-472 76432-474 76432-476 76432-478 76432-480 76432-482 76432-484 76432-486 76432-488 76432-490 76432-492 76432-494 76432-496 76432-498 76432-500 76432-502 76432-504 76432-506 76432-508 76432-510 76432-512 76432-514 76432-516 76432-518 76432-520 76432-522 76432-524 76432-526 76432-528 76432-530 76432-532 76432-534 76432-536 76432-538 76432-540 76432-542 76432-544 76432-546 76432-548 76432-550 76432-552 76432-554 76432-556 76432-558 76432-560 76432-562 76432-564 76432-566 76432-568 76432-570 76432-572 76432-574 76432-576 76432-578 76432-580 76432-582 76432-584 76432-586 76432-588 76432-590 76432-592 76432-594 76432-596 76432-598 76432-600 76432-602 76432-604 76432-606 76432-608 76432-610 76432-612 76432-614 76432-616 76432-618 76432-620 76432-622 76432-624 76432-626 76432-628 76432-630 76432-632 76432-634 76432-636 76433-942 76433-944 76433-946 76433-948 76433-950 76433-952 76433-954 76433-956 76433-958 76433-960 76433-962 76433-964 76433-966 76433-968 76433-970 76433-972 76433-974
76431-148EA 1437.42 USD
76431-148 76431-150 76431-152 76431-154 76431-156 76431-158 76431-160 76431-162 76431-164 76431-166 76431-168 76431-170 76432-398 76432-400 76432-402 76432-404 76432-406 76432-408 76432-410 76432-412 76432-414 76432-416 76432-418 76432-420 76432-422 76432-424 76432-426 76432-428 76432-430 76432-432 76432-434 76432-436 76432-438 76432-440 76432-442 76432-444 76432-446 76432-448 76432-450 76432-452 76432-454 76432-456 76432-458 76432-460 76432-462 76432-464 76432-466 76432-468 76432-470 76432-472 76432-474 76432-476 76432-478 76432-480 76432-482 76432-484 76432-486 76432-488 76432-490 76432-492 76432-494 76432-496 76432-498 76432-500 76432-502 76432-504 76432-506 76432-508 76432-510 76432-512 76432-514 76432-516 76432-518 76432-520 76432-522 76432-524 76432-526 76432-528 76432-530 76432-532 76432-534 76432-536 76432-538 76432-540 76432-542 76432-544 76432-546 76432-548 76432-550 76432-552 76432-554 76432-556 76432-558 76432-560 76432-562 76432-564 76432-566 76432-568 76432-570 76432-572 76432-574 76432-576 76432-578 76432-580 76432-582 76432-584 76432-586 76432-588 76432-590 76432-592 76432-594 76432-596 76432-598 76432-600 76432-602 76432-604 76432-606 76432-608 76432-610 76432-612 76432-614 76432-616 76432-618 76432-620 76432-622 76432-624 76432-626 76432-628 76432-630 76432-632 76432-634 76432-636 76433-942 76433-944 76433-946 76433-948 76433-950 76433-952 76433-954 76433-956 76433-958 76433-960 76433-962 76433-964 76433-966 76433-968 76433-970 76433-972 76433-974
Avantor® Hichrom HI-35 MS, GC Columns
Chromatography Columns
Proprietary low-polarity and low-bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and is also an excellent choice as a confirmation column for semivolatile environmental analysis.
  • Phase composition: Crossbonded low-polarity proprietary phase
  • Long lifetime
Max temperature limits may change depending on the film thickness.